Merck
  • Small-Molecule Modulation of TDP-43 Recruitment to Stress Granules Prevents Persistent TDP-43 Accumulation in ALS/FTD.
Small-Molecule Modulation of TDP-43 Recruitment to Stress Granules Prevents Persistent TDP-43 Accumulation in ALS/FTD.
Neuron (2019-07-06)
Mark Y Fang, Sebastian Markmiller, Anthony Q Vu, Ashkan Javaherian, William E Dowdle, Philippe Jolivet, Paul J Bushway, Nicholas A Castello, Ashmita Baral, Michelle Y Chan, Jeremy W Linsley, Drew Linsley, Mark Mercola, Steven Finkbeiner, Eric Lecuyer, Joseph W Lewcock, Gene W Yeo
ABSTRACT
Stress granules (SGs) form during cellular stress and are implicated in neurodegenerative diseases such as amyotrophic lateral sclerosis and frontotemporal dementia (ALS/FTD). To yield insights into the role of SGs in pathophysiology, we performed a high-content screen to identify small molecules that alter SG properties in proliferative cells and human iPSC-derived motor neurons (iPS-MNs). One major class of active molecules contained extended planar aromatic moieties, suggesting a potential to intercalate in nucleic acids. Accordingly, we show that several hit compounds can prevent the RNA-dependent recruitment of the ALS-associated RNA-binding proteins (RBPs) TDP-43, FUS, and HNRNPA2B1 into SGs. We further demonstrate that transient SG formation contributes to persistent accumulation of TDP-43 into cytoplasmic puncta and that our hit compounds can reduce this accumulation in iPS-MNs from ALS patients. We propose that compounds with planar moieties represent a promising starting point to develop small-molecule therapeutics for treating ALS/FTD.
Sigma-Aldrich
Purmorphamine, ≥98% (HPLC)
Sigma-Aldrich
Kenpaullone, ≥98%
Sigma-Aldrich
Tyrphostin A9, solid
Sigma-Aldrich
Dorsomorphin, ≥98% (HPLC)
Sigma-Aldrich
Y-27632 dihydrochloride, ≥98% (HPLC)
Sigma-Aldrich
DAPT, ≥98% (HPLC), solid
Sigma-Aldrich
CHIR99021, ≥98% (HPLC)
Sigma-Aldrich
Retinoic acid, ≥98% (HPLC), powder
Sigma-Aldrich
Penfluridol, ≥97% (HPLC), powder