Skip to Content
Merck
All Photos(1)

Documents

V900842

Sigma-Aldrich

Tris Acetate-EDTA buffer

Vetec, reagent grade, 10 ×, solution, RNase and DNase free

Synonym(s):

TAE buffer

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41105319
PubChem Substance ID:

grade

reagent grade

product line

Vetec

form

solution

feature

RNase and DNase free

concentration

10 ×

suitability

suitable for gel electrophoresis (after dilution to working concentration)

SMILES string

CC(O)=O.NC(CO)(CO)CO.OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O

InChI

1S/C10H16N2O8.C4H11NO3.C2H4O2/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20;5-4(1-6,2-7)3-8;1-2(3)4/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20);6-8H,1-3,5H2;1H3,(H,3,4)

InChI key

HGEVZDLYZYVYHD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

TAE running buffer is the most commonly used buffer for DNA agarose gel electrophoresis but is also used for non-denaturing RNA agarose gel electrophoresis. Double-stranded DNA tends to run faster in TAE than in other buffers but can also become exhausted during extended electrophoresis. Buffer circulation or buffer replacement during extended electrophoresis can remedy the lower buffering capacity. Dilution of the concentrated TAE buffer produces a 1× TAE buffer with 40 mM Tris-acetate and 1 mM EDTA, pH 8.3. The 1× TAE buffer is used both in the agarose gel and as a running buffer. Applied voltages of less than 5 V/cm (distance between the electrodes of the unit) are recommended for maximum resolution.

Preparation Note

Prepared with Biotechnology Performance Certified Trizma base (T6066) and Molecular Biology Reagent EDTA disodium salt (E5134). Solutions also contain acetic acid (A6283); powdered blend contains Trizma acetate (T1258).

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service