G1007243
HPLC Analysis of Tryptic Peptides from Ovalbumin on SeQuant™ ZIC-HILIC
application for HPLC
About This Item
technique(s)
HPLC: suitable
test parameters
column: SeQuant ZIC-HILIC, PEEK, 150 x 0.3 mm, 5 μm, 200 Å (1.50481.0001)
mobile phase: [A] 0.25% (v/v) formic acid with Milli-q-water; [B] 0.25% (v/v) formic acid in acetonitrile; (10:90, A:B)
gradient: Initial composition: 90% B and 10% A. Linear gradient from 0-40 min
flow rate: 5 μL/min
detector: MicroMass Ultima-QTOF, Needle voltage: 3.1 kV, Cone voltage: 45V (collision energy: 10V; Scan range: m/z 400-1800; 1.5 s cycle time)
injection: 1 μL of 10 pmole digested sample injected in mobile phase
suitability
application for HPLC
application(s)
life science and biopharma
Legal Information
Analyte
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service