Skip to Content
Merck
All Photos(1)

Documents

ERMDA477IFCC

Human serum (anti-ß2-glycoprotein I IgG)

certified reference material, ERM®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.77

grade

certified reference material

Agency

ERM®

manufacturer/tradename

JRC

application(s)

clinical testing

storage temp.

-65 to -96°C

Related Categories

Analysis Note

For more information please click on the below product number:
ERMDA477IFCC

Legal Information

ERM is a registered trademark of European Commission

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service