Skip to Content
Merck
All Photos(1)

Key Documents

CRM80884

Supelco

Diquat metabolite TOPPS

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

1-Oxo-1,2,3,4-tetrahydropyrido[1,2-a]pyrazin-5-ium bromide

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€119.00

€119.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
€119.00

About This Item

Empirical Formula (Hill Notation):
C8H9BrN2O
CAS Number:
Molecular Weight:
229.07
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€119.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

format

neat

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

Diquat metabolite TOPPS is a common degradation product of diquat. Diquat is a nonselective, defoliant, preharvest, and non-residual bipyridyl herbicide and crop desiccant, normally used as the dibromide salt, to control broad-leaved weeds and grasses. Its activity is based on the liberation of the superoxide anion radical and, subsequently, hydrogen peroxide, leading to tissue destruction by oxidative stress. The herbicidal activity of diquat is more pronounced in daylight, aerobic conditions, high humidity, and temperature.

As per Regulation (EC) No 1107/2009, repealing the directive 91/414/ECC, diquat is not approved for use in the European Union (EU). The maximum residue limit (MRL) for diquat presence in the crops varies between 0.01 mg/kg in most of the commodities to 5 mg/kg for linseed in accordance with Regulation (EU) 2016/1002.

Application

The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1 Oral - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service