Skip to Content
Merck
All Photos(1)

Key Documents

AR723

Supelco

Sulfur in Coke (S)

0.50% S, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107

grade

certified reference material

manufacturer/tradename

Alpha Resources

concentration

0.50% S

application(s)

petroleum

format

matrix material

Looking for similar products? Visit Product Comparison Guide

Other Notes

Certified for the analytes listed below. See certificate for values and more details
Elemental Analysis: Sulfur

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service