Skip to Content
Merck
All Photos(1)

Documents

89878

Supelco

1,1,1-Tris(hydroxymethyl)propane

analytical standard

Synonym(s):

2-Ethyl-2-hydroxymethyl-1,3-propanediol, Trimethylolpropane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2C(CH2OH)3
CAS Number:
Molecular Weight:
134.17
Beilstein:
1698309
MDL number:
UNSPSC Code:
41116107

grade

analytical standard

vapor density

4.8 (vs air)

vapor pressure

<1 mmHg ( 20 °C)

Assay

≥96.5% (GC)

form

powder

autoignition temp.

1301 °F

shelf life

limited shelf life, expiry date on the label

bp

159-161 °C/2 mmHg (lit.)

mp

56-58 °C (lit.)

SMILES string

CCC(CO)(CO)CO

InChI

1S/C6H14O3/c1-2-6(3-7,4-8)5-9/h7-9H,2-5H2,1H3

InChI key

ZJCCRDAZUWHFQH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food. Find all available reference materials for compounds listed in 10/2011 here

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

356.0 °F - Cleveland open cup

Flash Point(C)

180 °C - Cleveland open cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service