823428
TOYOPEARL® AF-rProtein A HC-650F Bulk Media
phase rProtein A, bottle of 1 L, 45 μm particle size
Synonym(s):
TOYOPEARL® AF-rProtein A HC-650F Bulk Media
About This Item
Recommended Products
description
Adsorption capacity (> 80 g/L IgG (static); >70 g/L IgG (dynamic at 5 min. residence time))
Clean in Place (CIP) (IIID clarifierID clarifier
ID clarif
ierID clarifier0.1)
Phase filter (Protein A)
product line
TOYOPEARL®
form
slurry
packaging
bottle of 1 L
manufacturer/tradename
Tosoh 23428
parameter
3 bar max. pressure
technique(s)
LPLC: suitable
matrix
hydroxylated methacrylic polymer particle platform
matrix active group
rProtein A phase
particle size
45 μm
pore size
1000 Å
pH
2-13
capacity
>70 g/L adsorption capacity (IgG)(dynamic at 5 min. residence time)
>80 g/L adsorption capacity (IgG)(static)
separation technique
affinity
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
General description
TOYOPEARL AF-rProtein A HC-650F resin maintains 80% of initial dynamic binding capacity after 40 CIP cycles with 0.5 mol/L NaOH. TOYOPEARL AF-rProtein A HC-650F resin exhibits dynamic binding capacities of greater than 65 g/L at residence times of 5 minutes and greater than 50 g/L at 2 minutes residence time with feed stock concentrations from 1.0 g/L to 10.0 g/L.
Achievement of high linear velocities at relatively low pressure enables high throughput at production scale using equipment with moderate pressure limitations. Improved mass transfer characteristics allow it to maintain a larger percent of its capacity at lower residence times relative to agarose base stable resins. Typical leakage for this rProtein A ligand is 0.6 -1.7 ng rProtein A /mg eluted antibody by ELISA testing. Though TOYOPEARL AF-rProtein A-650F resin is shipped in 20% ethanol, it can be stored in 2% benzyl alcohol if necessary.
Good pressure-flow characteristics and kinetics contribute to maintaining good capacity particularly at low residence times.
Specifications
Physical form
Legal Information
related product
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
103.1 °F
Flash Point(C)
39.5 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service