Skip to Content
Merck
All Photos(1)

Key Documents

48129

Supelco

Dibutyl chlorendate solution

200 μg/mL in acetonitrile, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H20Cl6O4
CAS Number:
Molecular Weight:
501.06
EC Number:
UNSPSC Code:
41121813

grade

analytical standard

feature

standard type surrogate

packaging

ampule of 1 mL

concentration

200 μg/mL in acetonitrile

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

compatibility

for analyte group Pesticides/PCBs

format

single component solution

storage temp.

2-8°C

InChI

1S/C17H20Cl6O4/c1-3-5-7-26-13(24)9-10(14(25)27-8-6-4-2)16(21)12(19)11(18)15(9,20)17(16,22)23/h9-10H,3-8H2,1-2H3

InChI key

UJAHPBDUQZFDLA-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service