Skip to Content
Merck
All Photos(1)

Key Documents

442292

Supelco

2,3-Dimethylphenol

analytical standard

Synonym(s):

3-Hydroxyl-o-xylene, vic.-o-Xylenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C6H3OH
CAS Number:
Molecular Weight:
122.16
Beilstein:
1906267
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

vapor density

4.23 (vs air)

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

217 °C (lit.)

mp

70-73 °C (lit.)

application(s)

agriculture
cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

room temp

SMILES string

Cc1cccc(O)c1C

InChI

1S/C8H10O/c1-6-4-3-5-8(9)7(6)2/h3-5,9H,1-2H3

InChI key

QWBBPBRQALCEIZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

203.0 °F - closed cup

Flash Point(C)

95 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Baggi et al.
Applied and environmental microbiology, 53(9), 2129-2132 (1987-09-01)
A Pseudomonas stutzeri strain capable of growing on o-xylene was isolated from enrichment cultures. The organism grew on 2,3- and 3,4-dimethylphenol but not on 2-methylbenzyl alcohol, o-tolualdehyde, or o-toluate. P. stutzeri was not able to utilize m-xylene, p-xylene, or 1,2,4-trimethylbenzene
A E Holliday et al.
Journal of chemical ecology, 38(3), 278-286 (2012-03-07)
The defensive secretion of the ground beetle Chlaenius cordicollis is predominantly 3-methylphenol. Adult C. cordicollis were collected in Pennsylvania and Manitoba and induced to discharge defensive secretion in a vial. The headspace was sampled by solid phase microextraction, and samples
Zulin Zhang et al.
Journal of separation science, 37(24), 3699-3705 (2014-10-02)
A simple and rapid method was developed for the simultaneous analysis of nine different pesticides in water samples by gas chromatography with mass spectrometry. A number of parameters that may affect the recovery of pesticides, such as the type of
Pingxi Xu et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(46), 16592-16597 (2014-10-29)
Insect repellents are important prophylactic tools for travelers and populations living in endemic areas of malaria, dengue, encephalitis, and other vector-borne diseases. DEET (N,N-diethyl-3-methylbenzamide) is a 6-decade-old synthetic repellent, which is still considered the gold standard of mosquito repellents. Mosquitoes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service