27010-U
Vials, screw top with phenolic open-top cap, pre-assembled
volume 40 mL, amber glass vial, O.D. × H 29 mm × 82 mm, tan PTFE/silicone septum
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
amber glass vial
black phenolic cap (open-top)
tan PTFE/silicone septum
feature
closure type black phenolic hole cap
closure type screw top vial
packaging
pkg of 100 ea
O.D. × H
29 mm × 82 mm
volume
40 mL
fitting
thread for 24-400
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Vials are suitable basically to inject samples from an autosampler. They are available in various sizes, although 2 mL vials are more commonly used. Autosampler needle pierces through the cap during injection and withdraws the required aliquots of sample from the vials. Amber glass vials (volume 40 mL, O.D. × H. 29 mm × 82 mm) are useful in preventing the sensitive analyte under investigation from exposure to UV light. The threads on the standard vials are 24-400. The phenolic cap is autoclavable and it can withstand the temperature up to 149° C. The tan PTFE/silicone septum is sized to fit screw top vials. It is suitable for use with most organics (until pierced) and is not recommended for use with chlorosilanes.
related product
replacement part
Product No.
Description
Pricing
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service