Key Documents
22116
Fine Metering Valve
new
brass, 1/8 in. straight arms
About This Item
UNSPSC Code:
41115717
eCl@ss:
32035302
Pricing and availability is not currently available.
material
brass
description
fine adjustment
fitting
1/8 in. straight arms
General description
Use for very accurate flow regulation. Install a flow measuring device downstream. Nuts and ferrules included. An optional vernier handle (Cat. No. 22122, order separately) is available for added convenience.
Application
Fine metering valve may be used on the exit of a flow system, which is basically used for the conductance measurements of decyl-, dodecyl-, tetradecyl- and hexadecyltrimethylammonium bromide in water at 25, 60, 95, 130, and 160°C, primarily done in order to determine their critical micelle concentrations (CMCs).[1]
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Paulami Bose et al.
Nanoscale, 12(43), 22116-22128 (2020-10-30)
Reactions between atomically precise noble metal nanoclusters (NCs) have been studied widely in the recent past, but such processes between NCs and plasmonic nanoparticles (NPs) have not been explored earlier. For the first time, we demonstrate spontaneous reactions between an
Critical micelle concentrations for alkyltrimethylammonium bromides in water from 25 to 160 C.
Fennell ED
Journal of Solution Chemistry, 13 (2), 87-101 (1984)