Skip to Content
Merck
All Photos(1)

Key Documents

13247-U

Supelco

Metal Packed GC Column (fits Agilent, Configuration "C")

phase none, matrix 50/80 Porapak Q support, L × O.D. × I.D. 8.0 ft (2.4 m) × 1/8 in. × 2.1 mm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115710
NACRES:
SB.54

material

stainless steel column

manufacturer/tradename

Agilent

parameter

0-250 °C temperature (isothermal or programmed)

technique(s)

gas chromatography (GC): suitable

L × O.D. × I.D.

8.0 ft (2.4 m) × 1/8 in. × 2.1 mm

matrix

50/80 Porapak Q support

matrix active group

none phase

column type

packed GC

compatibility

for use with Agilent Configuration A

Looking for similar products? Visit Product Comparison Guide

General description

An economical metal column should be used for less demanding applications where the inertness of glass is not required. This column fits Agilent 5880, 5890, 6820, 6890, 7820, and 7890 (configuration C, not-on-column injection, all detectors except TCD).
In packed columns the carrier gas percolates through the porous bed and separation is achieved by moving the solutes apart. Packed columns are made up of mainly two materials; stainless steel and Pyrex glass. Metal packed columns are more suitable for long and high carrier gas flow rates to tolerate the elevated pressures. Porapak Q is a porous copolymer of styrene and ethylvinylbenzene crosslinked with divinylbenzene which is applicable for quantitative adsorption.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The adsorption of enkephalin to porous polystyrene beads: a simple assay for enkephalin hydrolysis.
Z Vogel et al.
FEBS letters, 80(2), 332-336 (1977-08-15)
Raymond P. W. Scott
Introduction to Analytical Gas Chromatography (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service