Skip to Content
Merck
All Photos(1)

Documents

Z744321

Wilmad® Coaxial Insert

stem L 50 mm, reference capacity 200 µL, sample capacity 260 µL

Synonym(s):

Small Volume NMR, Reference NMR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121705
NACRES:
NB.15

packaging

pack of 1 ea

manufacturer/tradename

Wilmad WGS-5BL-SP

capacity

200 μL

stem L

50 mm

General description

The most versatile and reliable coaxial system available for various NMR experiments. General applications include small volume NMR, external referencing or locking and magnetic susceptibility determination. Ideal for variable temperature studies, all parts are made from Pyrex® glass. Note: Outer tube must be ordered separately based on magnetic field strength.

Application

NMR

Legal Information

PYREX is a registered trademark of Corning, Inc.
Wilmad is a registered trademark of SP Industries

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service