Key Documents
Z744320
Wilmad® 50 ml Suprasil® Small Volume Liquid Nitrogen Dewar
pack of 1
Synonym(s):
EPR cryo dewar, EPR dewar
Select a Size
24 EACH
€1,320.00
€1,320.00
Please contact Customer Service for Availability
Select a Size
Change View
24 EACH
€1,320.00
About This Item
UNSPSC Code:
41121804
NACRES:
NB.15
€1,320.00
Please contact Customer Service for Availability
packaging
pack of 1
manufacturer/tradename
Wilmad WG-816-Q
General description
Dewar Flasks provide a convenient alternative to Variable Temperature systems for studies at 77 K, and are silvered around the reservoir.
Legal Information
Suprasil is a registered trademark of HERAEUS QUARZGLAS GMBH & CO.
Wilmad is a registered trademark of SP Industries
Choose from one of the most recent versions:
Certificates of Analysis (COA)