Z744163
Wilmad® NMR Tube Carrier with Shirt Clip
to hold, NMR 1 tube, tube O.D. 5 mm, tube L 7 in., pack of 3
Synonym(s):
NMR tube carrier, NMR tube holder
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121705
Recommended Products
feature
to hold NMR 1 tube
packaging
pack of 3
manufacturer/tradename
Wilmad WG-6192
tube L
7 in.
tube O.D.
5 mm
General description
Handy pocket NMR/EPR tube carrier holds one 5mm O.D., 7 inch length NMR/EPR tube. Pack of 3 white holders. Features a pocket clip that keeps it secure. Made from unbreakable polymer material with a snap-on cap. Protection is provided for the sample and lab personnel during transport.
Application
NMR
Legal Information
Wilmad is a registered trademark of SP Industries
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service