Skip to Content
Merck
All Photos(1)

Documents

Z720518

Nunc® EZ Flip conical centrifuge tubes

50 mL conical tube, racked

Synonym(s):

centrifuge tubes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121703
NACRES:
NB.11

material

cap (leakproof cap)
conical bottom tube
polypropylene

sterility

sterile

feature

RCF 9,500 × g

packaging

pack of 25
case of 500

manufacturer/tradename

Nunc 362697

capacity

50 mL

color

clear

Looking for similar products? Visit Product Comparison Guide

General description

EZ Flip conical centrifuge tubes
Ergonomic design has a hinged-cap system for one-handed opening and closing. Integrated leakproof cap prevents cross contamination and lost closures. Clarified polypropylene combines chemical resistance and high speed tolerance. 15 mL and 50 mL versions are available White writing area and flat caps for sample identification. Printed graduations. Standard dimensions compatible with centrifuge rotors.

  • Hinged-cap system with ergonomic design for one-handed opening and closing
  • Integrated leakproof cap prevents cross contamination and lost closures
  • Clarified polypropylene combines chemical resistance and high speed tolerance
  • White writing area and flat caps for sample identification
  • Printed graduations
  • Standard dimensions compatible with centrifuge rotors

Legal Information

EZ Flip is a trademark of Thermo Fisher Scientific or its subsidiaries
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service