Key Documents
Z708801
Unicore Punch Kit
I.D. 1 mm, pack of 25
Synonym(s):
harris punch, harris uni-core, uni-core, whatman punch, Harris Uni-Core
About This Item
UNSPSC Code:
41100000
Pricing and availability is not currently available.
packaging
pack of 25
manufacturer/tradename
Whatman Article No. 29002515 (US reference)
Whatman WB100073
I.D.
1 mm
General description
An inexpensive disposable multi-purpose coring tool designed for simultaneous sample extraction, collection and storage in one operation. Suitably samples thin and soft substrates including filter paper, gels and dried blood on cards. Units can be autoclaved at 250°C for limited re-use.
Other Notes
Global SIAL customers (excluding Japan) can now order any product from the Cytiva Life Sciences consumables portfolio (prepack only) using GE′s product numbering system.
The legacy GE Whatman products that use the SIAL "Z" product numbering system will be discontinued and directly replaced with a product number using a "WHA" prefix plus the GE product code. For reference, the legacy "Z" product number is identified as a synonym on the "WHA" product number listing.
replaced by
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)