Z665622
Ohaus® Explorer® Pro balance
EP114C, AC/DC input 230 V AC
Synonym(s):
laboratory balance, weighing balance
All Photos(1)
About This Item
UNSPSC Code:
41111502
Recommended Products
feature
CE compliant
CSA approved
manufacturer/tradename
EP114C
availability
available only in EU
weighing capacity measuring range
110 g, resolution: 1 mg
platform diam.
90 mm
General description
The Explorer Pro Series of standard level balances was designed for simplicity and includes the industry′s easiest user interface for easy setup and operation; no training is required.
- Simple 2 key menu operation and navigation
- Text prompts for fast setup
- 7 Application modes plus below balance weighing
- Brilliant backlit high resolution dot matrix display
- RS232 port for easy communication with computer or printer
- GLP compliant with user selectable outputs
- 5 Language operational display text
- Protective in-use cover and security bracket included
- Industry leading warranty and support
Legal Information
Explorer is a registered trademark of Ohaus Corp.
Ohaus is a registered trademark of Ohaus Corp.
Choose from one of the most recent versions:
Certificates of Analysis (COA)