Documents
Z355542
Millipore
Filter membranes for microbiological analysis
pore size 0.45 μm (HA), S-Pak, Black with white grid surface
About This Item
UNSPSC Code:
41104900
NACRES:
NB.24
material
cellulose acetate/cellulose nitrate mixed esters filter
manufacturer/tradename
Millipore HABG047S6
Millipore HABG04S6
pore size
0.45 μm (HA)
General description
Filter type HA, 0.45 μm pore size, is designed to give complete retention and maximum recovery of total coliforms and fecal coliform bacteria. Complies with applicable U.S. EPA Standard methods and ASTM specifications for membrane filters used for drinking water analysis.
Filter type HC, 0.7 μm pore size, is designed for improved recovery of stressed fecal coliform organisms, especially those found in chlorinated effluents. The larger pore size permits faster filtration if the water sample has a higher particulate burden.
Both filters are cellulose acetate/cellulose nitrate mixed esters.
S-Kit includes filter membrane plus absorbent pad, sterile, individually sealed, packed in 100′s in dispenser tubes.
S-Pak includes filter membrane, sterile, individually sealed.