Key Documents
Z331422
Lenz® round bottom flasks
capacity 100 mL, joint: ST/NS 14/23
Select a Size
1 EA
€17.10
€17.10
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
€17.10
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
€17.10
Please contact Customer Service for Availability
material
borosilicate glass 3.3
round bottom flask
single-neck
Joint
joint: ST/NS14/23
Agency
according to DIN 12 348
sterility
non-sterile
feature
closure type Standard Taper Joint
packaging
case of 1
manufacturer/tradename
Lenz 3.0014.37
capacity
100 mL
Legal Information
Lenz is a registered trademark of Lenz GmbH
Choose from one of the most recent versions:
Certificates of Analysis (COA)