Z317322
Eppendorf® racks for microcentrifuge tubes
For 24 × 0.5 ml tubes
All Photos(1)
About This Item
Recommended Products
manufacturer/tradename
Eppendorf® 0030 123.107
availability
not available in USA and Canada
General description
- 24-place capacity
- Unique swivel connector to interlock multiple racks for front-to-back and side-to-side configurations
- Non-slip stand
- Numbered positions
- UV-resistant
- Autoclavable
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)