Skip to Content
Merck
All Photos(3)

Key Documents

WH0001345M1

Sigma-Aldrich

Monoclonal Anti-COX6C antibody produced in mouse

clone 4G4-2A8, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-cytochrome c oxidase subunit VIc

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€507.00

€507.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
€507.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€507.00


Please contact Customer Service for Availability

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4G4-2A8, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunofluorescence: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... COX6C(1345)

General description

Cytochrome c oxidase (COX), the terminal enzyme of the mitochondrial respiratory chain, catalyzes the electron transfer from reduced cytochrome c to oxygen. It is a heteromeric complex consisting of 3 catalytic subunits encoded by mitochondrial genes and multiple structural subunits encoded by nuclear genes. The mitochondrially-encoded subunits function in electron transfer, and the nuclear-encoded subunits may be involved in the regulation and assembly of the complex. This nuclear gene encodes subunit VIc, which has 77% amino acid sequence identity with mouse COX subunit VIc. This gene is up-regulated in prostate cancer cells. A pseudogene COX6CP1 has been found on chromosomes 16p12. (provided by RefSeq)

Immunogen

COX6C (AAH00187, 1 a.a. ~ 75 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MAPEVLPKPRMRGLLARRLRNHMAVAFVLSLGVAALYKFRVADQRKKAYADFYRNYDVMKDFEEMRKAGIFQSVK

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service