Key Documents
TTRT96P150W-EP
Eppendorf twin.tec® Real-Time PCR Plates 96
Skirted, PCR clean, volume 150 μL, white plate (white wells), pack of 25
About This Item
UNSPSC Code:
41106300
material:
polypropylene wells
white plate (white wells)
size:
96 well
volume:
150 μL
working volume:
150 μL
sterility:
non-sterile
feature:
skirt
PCR clean
Skirted
Product TTRT96P150W-EP is not currently sold in your country. Contact Technical Service
material
polypropylene wells
white plate (white wells)
sterility
non-sterile
feature
skirt
PCR clean
Skirted
packaging
pack of 25
manufacturer/tradename
Eppendorf® 951022015 NA Only
technique(s)
PCR: suitable
size
96 well
volume
150 μL
working volume
150 μL
Related Categories
General description
The white wells of Eppendorf’s twin.tec real-time PCR plates increase the reflection of fluorescent light in real-time PCR (qPCR) and other fluorescence assays. The increased fluorescence from these specialized qPCR plates improves the sensitivity of your assays, while also improving reproducibility and consistency of your results.
Features and Benefits
  • White wells for better reflection
  • High mechanical stability
  • Raised rims for effective sealing
  • Skirted and semi-skirted frames
  • Optimal heat transfer due to reduced wall thickness
  • Autoclavable (121 °C, 20 min)
  • PCR clean and Eppendorf Forensic DNA Grade s available
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
twin.tec is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)