Skip to Content
Merck
All Photos(1)

Key Documents

T9275

Sigma-Aldrich

Glyceryl trioleate

~95%

Synonym(s):

(9Z)9-Octadecenoic acid 1,2,3-propanetriyl ester, 1,2,3-Tri(cis-9-octadecenoyl)glycerol, Glycerol trioleate, Glycerol triolein, Oleic acid triglyceride, Oleic triglyceride, TG(18:1(9Z)/18:1(9Z)/18:1(9Z)), Triolein

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C17H33COOCH2)2CHOCOC17H33
CAS Number:
Molecular Weight:
885.43
Beilstein:
1718692
EC Number:
MDL number:
UNSPSC Code:
12352211

Assay

~95%

bp

235-240 °C/18 mmHg (lit.)

density

0.91 g/mL (lit.)

storage temp.

2-8°C

SMILES string

[H]C(COC(CCCCCCC/C=C\CCCCCCCC)=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O

InChI

1S/C57H104O6/c1-4-7-10-13-16-19-22-25-28-31-34-37-40-43-46-49-55(58)61-52-54(63-57(60)51-48-45-42-39-36-33-30-27-24-21-18-15-12-9-6-3)53-62-56(59)50-47-44-41-38-35-32-29-26-23-20-17-14-11-8-5-2/h25-30,54H,4-24,31-53H2,1-3H3/b28-25-,29-26-,30-27-

InChI key

PHYFQTYBJUILEZ-IUPFWZBJSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Sealed ampule.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service