Skip to Content
Merck
All Photos(1)

Key Documents

T7641

Sigma-Aldrich

Tetrazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CH2N4
CAS Number:
Molecular Weight:
70.05
Beilstein:
105799
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Quality Level

mp

154-158 °C (lit.)

SMILES string

c1nnn[nH]1

InChI

1S/CH2N4/c1-2-4-5-3-1/h1H,(H,2,3,4,5)

InChI key

KJUGUADJHNHALS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Not recommended for use in DNA synthesis.

Warning

Do not heat over 90°C (danger of explosion)

Pictograms

Exploding Bomb

Signal Word

Danger

Hazard Statements

Hazard Classifications

Self-react. A

Storage Class Code

1 - Explosive hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuko Yoshikawa et al.
Inorganic chemistry, 50(22), 11729-11735 (2011-10-19)
We investigated the effects of antitumor-active tetrazolato-bridged dinuclear platinum(II) complexes [{cis-Pt(NH(3))(2)}(2)(μ-OH)(μ-tetrazolato-N(1),N(2))](2+) (1) and [{cis-Pt(NH(3))(2)}(2)(μ-OH)(μ-tetrazolato-N(2),N(3))](2+) (2) on the higher-order structure of a large DNA molecule (T4 phage DNA, 166 kbp) in aqueous solution through single-molecule observation by fluorescence microscopy. Complexes 1
Duncan J Wardrop et al.
Organic letters, 14(6), 1548-1551 (2012-03-01)
The development of a mild, base-free method for the generation of alkylidenecarbenes is reported. Treatment of 5-hydroxyalkyl-1H-tetrazoles with carbodiimides generates products arising from the 1,2-rearrangement or [1,5]-C-H bond insertion of a putative alkylidenecarbene. Formation of this divalent intermediate is proposed
Mohamad Sabbah et al.
Bioorganic & medicinal chemistry, 20(15), 4727-4736 (2012-07-04)
New analogues of N-acyl-homoserine-lactone (AHL), in which the amide was replaced by a triazole or tetrazole ring, were prepared and tested for their activity as LuxR-dependent QS modulators. Several compounds showed a level of antagonistic or agonistic activity, notably some
Zhipeng Yu et al.
Journal of the American Chemical Society, 133(31), 11912-11915 (2011-07-09)
Photoactivatable fluorescent probes are invaluable tools for the study of biological processes with high resolution in space and time. Numerous strategies have been developed in generating photoactivatable fluorescent probes, most of which rely on the photo-"uncaging" and photoisomerization reactions. To
Adina Morozan et al.
ChemSusChem, 5(4), 647-651 (2012-03-06)
High-performance oxygen reduction reaction (ORR) catalysts based on metal-free nitrogen-containing precursors and carbon nanotubes are reported. The investigated systems allow the evaluation of the effect of nitrogen-containing groups towards ORR and the results show that the catalysts are compatible with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service