Skip to Content
Merck
All Photos(1)

Key Documents

T4389

Sigma-Aldrich

(+)-α-Tocopherol

from vegetable oil, Type VI, liquid, ≥600 IU/g

Synonym(s):

2,5,7,8-Tetramethyl-2-(4′,8′,12′-trimethyltridecyl)-6-chromanol, 5,7,8-Trimethyltocol, D-α-Tocopherol, Vitamin E

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H50O2
CAS Number:
Molecular Weight:
430.71
Beilstein:
4712525
EC Number:
MDL number:
UNSPSC Code:
12352200

biological source

vegetable oil

type

Type VI

form

liquid

specific activity

≥600 IU/g

color

clear yellow

refractive index

n20/D 1.505 (lit.)

bp

200-220 °C/0.1 mmHg (lit.)

density

0.95 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCc2c(C)c(O)c(C)c(C)c2O1

InChI

1S/C29H50O2/c1-20(2)12-9-13-21(3)14-10-15-22(4)16-11-18-29(8)19-17-26-25(7)27(30)23(5)24(6)28(26)31-29/h20-22,30H,9-19H2,1-8H3/t21-,22-,29-/m1/s1

InChI key

GVJHHUAWPYXKBD-IEOSBIPESA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Mixed isomers

Quality

Approx. 400 mg of D-α-tocopherol per gram.
The non-α-content varies from 5-50 mg per gram, the remainder is soybean oil.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service