Skip to Content
Merck
All Photos(1)

Documents

T301000

Sigma-Aldrich

DMT-dT-CPG

pore size 500 Å, 30-40μmol/g

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.51

type

for DNA synthesis

product line

Proligo Reagents

Assay

≥98.0% (Nucleoside Purity, reversed phase HPLC)

pore size

500 Å

capacity

≥99 % (Coupling Efficiency)

λ

conforms (UV/VIS Identity)

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service