Skip to Content
Merck
All Photos(1)

Key Documents

T1535

Sigma-Aldrich

Trizma® hydrochloride

≥99% (redox titration)

Synonym(s):

TRIS HCl, TRIS hydrochloride, Tris(hydroxymethyl)aminomethane hydrochloride, Tromethane hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2C(CH2OH)3 · HCl
CAS Number:
Molecular Weight:
157.60
Beilstein:
3675235
EC Number:
MDL number:
UNSPSC Code:
12161700

Assay

≥99% (redox titration)

impurities

≤0.5% water (Karl Fischer)

useful pH range

7.0-9.0

pKa (25 °C)

8.1

solubility

40 % (w/w), clear, colorless

cation traces

heavy metals (as Pb): ≤0.0005%

absorption

≤0.05 at 290 at 40%

SMILES string

Cl.NC(CO)(CO)CO

InChI

1S/C4H11NO3.ClH/c5-4(1-6,2-7)3-8;/h6-8H,1-3,5H2;1H

InChI key

QKNYBSVHEMOAJP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

The pH values of all buffers are temperature and concentration dependent. For Tris buffers, pH increases about 0.03 unit per °C decrease in temperature, and decreases 0.03-0.05 unit per ten-fold dilution. For precise applications, use a carefully calibrated pH meter with a glass/calomel combination electrode.

Legal Information

Trizma is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Ranjit et al.
Vascular pharmacology, 71, 102-107 (2015-04-15)
Research and routine laboratory assessment of clot integrity can be time consuming, expensive, and cannot be batched as it is generally performed in real time. To address these issues, we developed and validated a micro-titre based assay to quantify thrombogenesis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service