Skip to Content
Merck
All Photos(1)

Key Documents

SRP6258

Sigma-Aldrich

Granzyme B human

recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)

Synonym(s):

CCPI, CGL-1, CSP-B, CSPB, CTLA1, CTSGL1, Fragmentin-2, GRB, GZMB, Granzyme B, Granzyme-2, HLP, SECT

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in HEK 293 cells

tag

6-His tagged (C-terminus)

Assay

≥95% (SDS-PAGE)

form

lyophilized powder

mol wt

calculated mol wt 26.4 kDa
observed mol wt 35 kDa (DTT-reduced. Protein migrates due to glycosylation. Ile 21 is the predicted N-terminus.)

packaging

pkg of 10 μg

impurities

<1 EU/μg endotoxin (LAL test)

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... GZMB(3002)

General description

Granzyme B (GZMB) also known as Cathepsin G-like 1 (CTSGL1), Cytotoxic T-lymphocyte proteinase 2 (Lymphocyte protease), Fragmentin-2, Granzyme-2, Human lymphocyte protein (HLP), SECT, CGL1, CSPB, CTLA1, GRB, which belongs to the peptidase S1 family. Granzyme subfamily GZMB / CTSGL1 contains 1 peptidase S1 domain. The catalytic activity of Granzyme B is “Preferential cleavage: -Asp-|-Xaa- >> -Asn-|-Xaa- > -Met-|-Xaa-, -Ser-|-Xaa-“, and is inactivated by the serine protease inhibitor diisopropyl fluorophosphates. GZMB is necessary for target cell lysis in cell-mediated immune responses. It cleaves after Asp. GZMB seems to be linked to activation cascade of caspases (aspartate-specific cysteine proteases) responsible for apoptosis execution and cleaves caspase-3, -7, -9 and 10 to give rise to active enzymes mediating apoptosis.

Physical form

Lyophilized from 0.22 μm filtered solution in PBS. Generally 5-8% Mannitol or trehalose is added as a protectant before lyophilization.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in sterile PBS, pH 7.4 to a concentration of 50 μg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 month. For extended storage, it is recommended to store at -20°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service