Skip to Content
Merck
All Photos(1)

Key Documents

SRP3235

Sigma-Aldrich

SF-20 from mouse

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Synonym(s):

IL-25

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€346.00

€346.00


Please contact Customer Service for Availability


Select a Size

Change View
10 μG
€346.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

€346.00


Please contact Customer Service for Availability

biological source

mouse

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

mol wt

15.8 kDa

packaging

pkg of 10 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

General description

Murine SF20 is a bone marrow stroma-derived growth factor. SF20 is expressed in bone marrow, spleen stroma cells, resting mononuclear cells, resting CD8+ and CD19+ cells and activated CD8+ T cells and has been shown to bind to the surface of cells expressing the receptor TSA-1 (Thymic shared Ag-1). Among its biological activities, SF20 stimulates the proliferation of FDCP2 cells (a mouse factor-dependent hemopoietic cell line) and mouse lymphoid cells. Recombinant murine SF20 is a 15.8 kDa protein containing 143 amino acid residues.

Biochem/physiol Actions

Murine SF20 is a bone marrow stroma-derived growth factor. Recombinant murine SF20 is a 15.8 kDa protein containing 143 amino acid residues.

Sequence

MVSEPTTVPF DVRPGGVVHS FSQDVGPGNK FTCTFTYASQ GGTNEQWQMS LGTSEDSQHF TCTIWRPQGK SYLYFTQFKA ELRGAEIEYA MAYSKAAFER ESDVPLKSEE FEVTKTAVSH RPGAFKAELS KLVIVAKAAR SEL

Physical form

Lyophilized with no additives.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service