Skip to Content
Merck
All Photos(1)

Key Documents

SRP3163

Sigma-Aldrich

sTNF-receptor Type II human

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Synonym(s):

Soluble Tumor Necrosis Factor type II, TNFR75, TNFRSF1B, p75

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
€250.00

€250.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
20 μG
€250.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

€250.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

human

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

potency

0.25 ng/mL ED50

mol wt

18.9 kDa

packaging

pkg of 20 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... TNFRSF1B(7133)

General description

TNFRII is a member of the TNFR superfamily of transmembrane proteins, and is expressed in immune cells and certain endothelial cells. It is a high affinity receptor for TNF-a but manifests a lower affinity to TNF-b. Signaling through this receptor regulates various biological processes including cell proliferation, differentiation, apoptosis, lipid metabolism, coagulation, and neurotransmission. Soluble TNFRII is capable of inhibiting TNF-a induced activities by acting as a decoy receptor. Recombinant human sTNFRII is an 18.9 kDa protein (174 amino acid residues) comprising the cysteine rich ligand binding portion of the extracellular domain of the TNFRII protein.

Biochem/physiol Actions

TNFRII is a member of the TNFR superfamily of transmembrane proteins, and is expressed in immune cells and certain endothelial cells. Recombinant human sTNFRII is an 18.9 kDa protein (174 amino acid residues) comprising the cysteine rich ligand binding portion of the extracellular domain of the TNFRII protein.

Sequence

MAPEPGSTCR LREYYDQTAQ MCCSKCSPGQ HAKVFCTKTS DTVCDSCEDS TYTQLWNWVP ECLSCGSRCS SDQVETQACT REQNRICTCR PGWYCALSKQ EGCRLCAPLR KCRPGFGVAR PGTETSDVVC KPCAPGTFSN TTSSTDICRP HQICNVVAIP GNASMDAVCT STSP

Physical form

Lyophilized from 10 mM Sodium Phosphate, pH 8.0 + 50 mM NaCl.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aggarwal, B.B., et al.
Cytokine Reference, 1619-1632 (2001)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service