Skip to Content
Merck
All Photos(1)

Documents

SRP0677

Sigma-Aldrich

S6K Substrate

≥95% (HPLC), lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

product name

S6K Substrate, ≥95% (HPLC)

Assay

≥95% (HPLC)

form

lyophilized powder

mol wt

1155.43

shipped in

ambient

storage temp.

−20°C

Application

Useful as a substrate for active protein kinases, including CLK2, MRCKalpha, MRCKbeta, p70S6K, ROCK1, ROCK2, RSK1, RSK3, PIM1 and PIM2.

Biochem/physiol Actions

The S6K sub peptide sequence (KRRRLASLR) is based on human 40S ribosomal protein S6 (amino acid 230-238).

Sequence

KRRRLASLR

Reconstitution

Reconstitute in 20mM Tris-HCl, pH 7.5 solution to a final concentration of 1mg/mL.

Storage and Stability

Store diluted product in aliquots at -20°C. Avoid freeze/thaw cycles. Stability: >1yr at -20°C

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service