Skip to Content
Merck
All Photos(1)

Key Documents

SRP0479

Sigma-Aldrich

GCN5 (727-837) human

recombinant, expressed in E. coli, ≥90% (SDS-PAGE)

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€734.00

€734.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
€734.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

€734.00


Please contact Customer Service for Availability

biological source

human

recombinant

expressed in E. coli

Assay

≥90% (SDS-PAGE)

form

aqueous solution

mol wt

14.1 kDa

packaging

pkg of 100 μg

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... KAT2A(2648)

General description

Human GCN5 or KAT2A containing bromodomain (GenBank Accession No. NM_021078) amino-acids 727 – 837 (end) with N-terminal HIS-tag, MW = 14.1 kDa, expressed in an E. coli expression system.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shanshan Li et al.
The Journal of biological chemistry, 284(14), 9411-9417 (2009-02-17)
Bromodomains are acetyl lysine binding modules found in many complexes that regulate gene transcription. In budding yeast, the coactivator complex SAGA (Spt-Ada-Gcn5-acetyl-transferase) predominantly facilitates transcription of stress-activated genes and requires the bromodomain of the Gcn5 subunit for full activation of
Donna A Santillan et al.
Cancer research, 66(20), 10032-10039 (2006-10-19)
A critical unanswered question about mixed lineage leukemia (MLL) is how specific MLL fusion partners control leukemia phenotype. The MLL-cyclic AMP-responsive element binding protein-binding protein (CBP) fusion requires both the CBP bromodomain and histone acetyltransferase (HAT) domain for transformation and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service