Skip to Content
Merck
All Photos(2)

Key Documents

SRP0454

Sigma-Aldrich

BRD2, BD1, BD2 (65-459) GST tag human

recombinant, expressed in E. coli, ≥80% (SDS-PAGE)

Synonym(s):

FSH, NAT, RING3, RNF3, bromodomain containing 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€541.00

€541.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
€541.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

€541.00


Please contact Customer Service for Availability

biological source

human

recombinant

expressed in E. coli

Assay

≥80% (SDS-PAGE)

form

aqueous solution

mol wt

71.1 kDa

packaging

pkg of 100 μg

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... BRD2(6046)

General description

Human Bromodomain containing 2, also known as BRD2, RING3 or RNF3 (GenBank Accession No. NM_005104) amino-acids 65 – 459 with N-terminal GST-tag, MW = 71.1 kDa, expressed in an E. coli expression system.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takashi Umehara et al.
FEBS letters, 584(18), 3901-3908 (2010-08-17)
The BET family proteins recognize acetylated chromatin through their two bromodomains, acting as transcriptional activators or tethering viral genomes to the mitotic chromosomes of their host. The structural mechanism for how the N-terminal bromodomain of human BRD2 (BRD2-BD1) deciphers the
Enyuan Shang et al.
Journal of cellular biochemistry, 112(10), 2784-2793 (2011-05-25)
The human BRD2 gene has been linked and associated with a form of common epilepsy and electroencephalographic abnormalities. Disruption of Brd2 in the mouse revealed that it is essential for embryonic neural development and that viable Brd2(+/-) heterozygotes show both

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service