Skip to Content
Merck
All Photos(1)

Documents

SML3491

Sigma-Aldrich

Enflicoxib

≥98% (HPLC)

Synonym(s):

(±)-E 6087, 1-(4-aminosulphonylphenyl)-5-(2,4-difluorophenyl)-4,5-dihydro-3-trifluoromethyl-1H-pyrazole, 4-(5-(2,4-difluorophenyl)-3-(trifluoromethyl)-4,5-dihydro-1H-pyrazol-1-yl)benzenesulfonamide, E-6087

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H12F5N3O2S
CAS Number:
Molecular Weight:
405.34
UNSPSC Code:
51111800
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

O=S(N)(C1=CC=C(N2N=C(C(F)(F)F)CC2C3=CC=C(C=C3F)F)C=C1)=O

Biochem/physiol Actions

Orally available, potent and selective cyclooxygenase 2 (COX-2) inhibitor

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - STOT RE 2

Target Organs

Gastrointestinal tract

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service