Skip to Content
Merck
All Photos(1)

Key Documents

SML2250

Sigma-Aldrich

BB-Cl-amidine trifluoroacetate salt

≥98% (HPLC)

Synonym(s):

N-[(1S)-1-(1H-Benzimidazol-2-yl)-4-[(2-chloro-1-iminoethyl)amino]butyl]-[1,1′-biphenyl]-4-carboxamide trifluoroacetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H26ClN5O · xC2HF3O2
CAS Number:
Molecular Weight:
459.97 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77

Pricing and availability is not currently available.

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

O=C(C1=CC=C(C2=CC=CC=C2)C=C1)N[C@H](C3=NC4=C(N3)C=CC=C4)CCCNC(CCl)=N

Biochem/physiol Actions

BB-Cl-amidine is an irreversible pan-protein-arginine deiminase (PAD) inhibitor (kinact/KI in M-1min-1 = 16100/PAD1, 4100/PAD2, 6800/PAD3, 13300/PAD4) with >20-fold increased potency against U2OS osteosarcoma cutlures (viability assay IC50 = 8.8 μM vs >200 μM with Cl-amidine) and longer in vivo half-life (1.75 hrs vs 15 min for Cl-amidine in plasma post injection in mice) than Cl-amidine. BB-Cl-amidine effectively inhibits PMA-induced neutrophils extracellular trap (NET) formation from lupus-prone MRL/lpr mice-derived bone marrow neutrophils in cultures (by 78% and 34% with 20 μM BB-Cl-amidine or Cl-amidine, respectively), while daily s.c. injection (1 mg/kg) is efficacious in ameliorating lupus symptoms in in MRL/lpr mice in vivo.
Irreversible pan-protein-arginine deiminase (PAD) inhibitor with >10-fold increased potency than Cl-amidine both in vitro and in vivo.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fatemeh Ghari et al.
Science advances, 2(2), e1501257-e1501257 (2016-03-19)
Peptidyl arginine deiminase 4 (PAD4) is a nuclear enzyme that converts arginine residues to citrulline. Although increasingly implicated in inflammatory disease and cancer, the mechanism of action of PAD4 and its functionally relevant pathways remains unclear. E2F transcription factors are
Daniel M Lewallen et al.
ACS chemical biology, 10(11), 2520-2528 (2015-09-12)
Anti-citrullinated protein antibodies (ACPAs) are a hallmark of rheumatoid arthritis (RA) and are routinely used for disease diagnosis. Protein citrullination is also increased in cancer and other autoimmune disorders, suggesting that citrullinated proteins may serve as biomarkers for diseases beyond

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service