Skip to Content
Merck
All Photos(1)

Key Documents

SML2214

Sigma-Aldrich

BI01383298

≥98% (HPLC)

Synonym(s):

1-(3,5-Dichlorophenylsulfonyl)-N-(4-fluorobenzyl)piperidine-4-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H19Cl2FN2O3S
CAS Number:
Molecular Weight:
445.34
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

ClC1=CC(S(N2CCC(C(NCC3=CC=C(F)C=C3)=O)CC2)(=O)=O)=CC(Cl)=C1

Biochem/physiol Actions

BI01383298 is a selective probe for the Na(+)/citrate co-transporter (NaCT), SLC13A5. For characterization details of BI01383298, please visit the BI01383298 probe summary on the Structural Genomics Consortium (SGC) website.

To learn about other SGC chemical probes, visit sigma.com/sgc

Features and Benefits

BI01383298 is a chemical probe available through a partnership with the Structural Genomics Consortium (SGC). To learn more and view other SGC chemical probes, visit sigma.com/SGC.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service