Skip to Content
Merck
All Photos(1)

Key Documents

SML1738

Sigma-Aldrich

RL648_81

≥98% (HPLC)

Synonym(s):

Ethyl (2-amino-3-fluoro-4-((4-(trifluoromethyl)benzyl)amino)phenyl)carbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H17F4N3O2
CAS Number:
Molecular Weight:
371.33
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 25 mg/mL, clear

storage temp.

room temp

SMILES string

NC1=C(NC(OCC)=O)C=CC(NCC2=CC=C(C(F)(F)F)C=C2)=C1F

Biochem/physiol Actions

Ethyl (2-amino-3-fluoro-4-((4-(trifluoromethyl)benzyl)amino)phenyl)carbamate (RL648_81) is 15 times more effective and more selective than retigabine, an anticonvulsant. RL648_81 helps to treat or inhibit neurologic disorders linked with neuronal hyperexcitability.
RL648_81 is a selective and potent KCNQ2/3 channel activator.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and evaluation of potent KCNQ2/3-specific channel activators.
Kumar M, et al.
Molecular Pharmacology, 89(6), 667-677 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service