Skip to Content
Merck
All Photos(1)

Documents

SML1315

Sigma-Aldrich

Ranirestat

≥97% (HPLC)

Synonym(s):

(3R)-2′-[(4-Bromo-2-fluorophenyl)methyl]-spiro[pyrrolidine-3,4′(1′H)-pyrrolo[1,2-a]pyrazine]-1′,2,3′,5(2′H)-tetrone., AS-3201

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H11BrFN3O4
CAS Number:
Molecular Weight:
420.19
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥97% (HPLC)

form

powder

optical activity

[α]/D -5 to -10°, c = 1.0 in methanol

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

−20°C

SMILES string

O=C(N1CC2=C(F)C=C(Br)C=C2)C3=CC=CN3[C@@]4(CC(NC4=O)=O)C1=O

InChI

1S/C17H11BrFN3O4/c18-10-4-3-9(11(19)6-10)8-21-14(24)12-2-1-5-22(12)17(16(21)26)7-13(23)20-15(17)25/h1-6H,7-8H2,(H,20,23,25)/t17-/m1/s1

InChI key

QCVNMNYRNIMDKV-QGZVFWFLSA-N

Related Categories

Biochem/physiol Actions

Ranirestat (AS-3201) is an aldose reductase (AR) inhibitor. Aldose reductase is a rate-limiting enzyme in the conversion of D-glucose to fructose via D-sorbitol. Ranirestat has been implicated in diabetic complications (cataract formation, retinopathy, neuropathy, nephropathy). Ranirestat prevents sorbitol accumulation and is thus hoped to be effective in the remedy of diabetic neuropathy.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service