Skip to Content
Merck
All Photos(1)

Key Documents

SML1188

Sigma-Aldrich

PYZD-4409

≥98% (HPLC)

Synonym(s):

1-(3-Chloro-4-fluorophenyl)-4-[(5-nitro-2-furanyl)methylene]-3,5-pyrazolidinedione

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€89.30

€89.30


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€89.30

About This Item

Empirical Formula (Hill Notation):
C14H7ClFN3O5
CAS Number:
Molecular Weight:
351.67
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€89.30


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

, white to very dark brown

solubility

DMSO: 10 mg/mL, clear

storage temp.

−20°C

SMILES string

FC1=CC=C(N2C(C(C(N2)=O)=CC3=CC=C([N+]([O-])=O)O3)=O)C=C1Cl

InChI

1S/C14H7ClFN3O5/c15-10-5-7(1-3-11(10)16)18-14(21)9(13(20)17-18)6-8-2-4-12(24-8)19(22)23/h1-6H,(H,17,20)

InChI key

MSYMKEYWUWVZQY-UHFFFAOYSA-N

Biochem/physiol Actions

PYZD-4409 inhibts ATP-dependent activation of human ubiquitin activating enzyme E1 with an IC50 of 20 mM. PYZD-4409 induces cell death, and inhbits clonogenic growth of leukemia and myeloma cell lines, and inhibits leukemia xenograft tumor growth in mice.
PYZD-4409 inhibts ATP-dependent activation of human ubiquitin activating enzyme E1.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service