Skip to Content
Merck
All Photos(17)

Documents

SAB5700360

Sigma-Aldrich

Anti-Phospho-ERK1-T202/Y204 + ERK2-T185/Y187 antibody produced in rabbit

Synonym(s):

MAPK1/MAPK3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

36
38
40
41
43

species reactivity

rat, human, mouse

concentration

1.02 mg/ml

technique(s)

immunohistochemistry: 1:50-1:100
immunoprecipitation (IP): 1:50-1:100
western blot: 1:500-1:2000

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation ((pThr202/pTyr204)/(pThr185/pTyr187))

Gene Information

human ... MAPK1(5594)

Immunogen

A synthetic phosphorylated peptide around T185/Y187 of human ERK2 (NP_002736.3).

Physical form

PBS with 0.02% sodium azide,50% glycerol,pH7.3.

Storage and Stability

Store at -20℃. Avoid freeze / thaw cycles.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service