Skip to Content
Merck
All Photos(1)

Key Documents

SAB4700774

Sigma-Aldrich

Monoclonal Anti-Acrosin antibody produced in mouse

clone ACR-2, purified immunoglobulin, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

ACR-2, monoclonal

form

buffered aqueous solution

species reactivity

porcine

concentration

1 mg/mL

technique(s)

immunoblotting: suitable
immunocytochemistry: suitable

isotype

IgG1

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

pig ... Acr(397098)

General description

The antibody ACR-2 reacts with various forms of porcine acrosin (55, 53, 45 and 35 kDa), a typical serine proteinase with trypsin-like specificity. Acrosin is stored in the acrosome of undamaged spermatozoa.

Immunogen

Acid extracts of boar spermatozoa were subjected to hydrophobic chromatography and the pooled fraction with reactivity to N-alpha benzoylarginine-4-nitroanilide was used for immunization.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4, with 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service