Skip to Content
Merck
All Photos(1)

Key Documents

S8946

Sigma-Aldrich

Saikosaponin A from Bupleurum falcatnum

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H68O13
Molecular Weight:
780.98
UNSPSC Code:
12352200
PubChem Substance ID:

storage temp.

2-8°C

SMILES string

C[C@H]1O[C@@H](O[C@H]2CC[C@@]3(C)[C@@H](CC[C@]4(C)[C@@H]3C=C[C@]56OC[C@@]7(CCC(C)(C)C[C@@H]57)[C@@H](O)C[C@@]46C)[C@]2(C)CO)[C@H](O)[C@@H](O[C@@H]8O[C@H](CO)[C@@H](O)[C@H](O)[C@H]8O)[C@H]1O

InChI

1S/C42H68O13/c1-21-28(46)33(55-34-31(49)30(48)29(47)22(18-43)53-34)32(50)35(52-21)54-27-10-11-37(4)23(38(27,5)19-44)8-12-39(6)24(37)9-13-42-25-16-36(2,3)14-15-41(25,20-51-42)26(45)17-40(39,42)7/h9,13,21-35,43-50H,8,10-12,14-20H2,1-7H3/t21-,22-,23-,24-,25-,26+,27+,28+,29-,30+,31-,32-,33+,34+,35+,37+,38+,39-,40+,41-,42+/m1/s1

InChI key

KYWSCMDFVARMPN-MSSMMRRTSA-N

Application

Saikosaponins such as Saikosaponin A are used to help analyse and resolve the activities of herbal medicines such as Sho-saiko-to.

Biochem/physiol Actions

Saikosaponin A, a triterpenoid glycoside, induces apoptotic mechansims in human breast cancer cell lines. Saikosaponin A also displays an inhibitory activity against allergic asthma.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sho-saiko-to: Japanese herbal medicine for protection against hepatic fibrosis and carcinoma.
Shimizu I.
Journal of Gastroenterology and Hepatology, 15, Suppl D84-Suppl D90 (2000)
I Sakaida et al.
Journal of hepatology, 28(2), 298-306 (1998-03-26)
A herbal medicine, Sho-saiko-to (TJ-9), has recently been orally administered to patients with chronic liver disease in Japan and has been reported to inhibit the development of hepatocellular carcinoma. The aim of this study was to investigate whether TJ-9 has
K Kayano et al.
Journal of hepatology, 29(4), 642-649 (1998-11-21)
It is of extreme importance to prevent liver fibrosis and subsequent progression to liver cirrhosis. The aim of our study was to elucidate in vitro whether Sho-saiko-to exerted inhibitory effects on hepatic stellate cells. Hepatic stellate cells were isolated from
Yinhong Zhu et al.
International immunopharmacology, 72, 131-137 (2019-04-14)
Saikosaponin a (SSa), one of the major active components of Bupleurum falcatum, has antioxidant and anti-inflammatory pharmacological properties. However, the effects of SSa on liver injury have not been reported. In the present study, we evaluated the protective effects and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service