Recommended Products
biological source
rat lung
packaging
pkg of 500,000 cells
manufacturer/tradename
Cell Applications, Inc
growth mode
Adherent
morphology
fibroblast
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
asthma; tuberculosis; cancer; cardiovascular diseases
shipped in
dry ice
storage temp.
−196°C
General description
Lot specific orders are not able to be placed through the web. Contact your local sales rep for more details.
RLF from Cell Applications, Inc. have been used to demonstrate that exaggerated (compared to adults) NF-κB activation in neonates in response to hyperoxia is mediated by phosphorylation of IκBα on Tyr42 (Wright, 2009). They were also used to elucidate melatonin effects and signaling pathway (Ben Soussia, 2012) and served as a control in studies investigating epithelial-mesenchimal transition as the source of myofibroblasts responsible for idiopathic pulmonary fibrosis (Willis, 2005; Jain, 2007).
RLF from Cell Applications, Inc. have been used to demonstrate that exaggerated (compared to adults) NF-κB activation in neonates in response to hyperoxia is mediated by phosphorylation of IκBα on Tyr42 (Wright, 2009). They were also used to elucidate melatonin effects and signaling pathway (Ben Soussia, 2012) and served as a control in studies investigating epithelial-mesenchimal transition as the source of myofibroblasts responsible for idiopathic pulmonary fibrosis (Willis, 2005; Jain, 2007).
Cell Line Origin
Lungs
Application
Pulmonary function, gene expression, healing & repair process, differentiation, extracellular matrix production, growth factor and cytokine activation, actin reorganization, synthesis of type II collagen, elastin, and proteoglycans, airway remodeling, pharmaceutical development.
Components
Rat Fibroblast Basal Medium that contains 10% FBS and 10% DMSO
Quality
Each lot was tested for proper morphology, Population Doublings, Negative for HIV, Hepatitis B, Hepatitis C, mycoplasma, bacteria, and fungi.
Preparation Note
- 2nd passage, >500,000 cells in Rat Fibroblast Basal Medium that contains 10% FBS and 10% DMSO
- Can be cultured at least 10 population doublings
Subculture Routine
Please refer to the RLF Culture Protocol.
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service