Skip to Content
Merck
All Photos(1)

Documents

PZ0100

Sigma-Aldrich

CP-154526 hydrochloride

≥98% (HPLC)

Synonym(s):

N-Butyl-N-ethyl-2,5-dimethyl-7-(2,4,6-trimethylphenyl)-7H-pyrrolo[2,3-d]pyrimidin-4-amine monohydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H32N4 · HCl
CAS Number:
Molecular Weight:
400.99
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

Cl.CCCCN(CC)c1nc(C)nc2n(cc(C)c12)-c3c(C)cc(C)cc3C

InChI

1S/C23H32N4.ClH/c1-8-10-11-26(9-2)22-20-18(6)14-27(23(20)25-19(7)24-22)21-16(4)12-15(3)13-17(21)5;/h12-14H,8-11H2,1-7H3;1H

InChI key

ZWDLBNCJWNENFF-UHFFFAOYSA-N

Biochem/physiol Actions

CP-154526 is a selective, non-peptide antagonist of corticotropin releasing factor receptors (CRF1).

Features and Benefits

This compound is featured on the Corticotropin-Releasing Factor Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service