Skip to Content
Merck
All Photos(2)

Key Documents

PLA0199

Sigma-Aldrich

Anti-Vimentin Antibody

from rabbit

Synonym(s):

CTRCT30, HEL113, epididymis luminal protein 113

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

Product Name

Rabbit anti-Vimentin Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

grade

Powered by Bethyl Laboratories, Inc.

species reactivity

mouse, human

technique(s)

immunohistochemistry: 1:200-1:1,000
western blot: 1:2,000- 1:10,000

accession no.

NP_003371.2

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

rabbit ... Vimentin(7431)

General description

Vimentin is a specific type III intermediate filament (IF) protein. It is abundantly expressed in mesenchymal cells. The gene encoding this protein is localized on human chromosome 10p13.

Immunogen

The epitope recognized by PLA0199 maps to a region between residue 416 and 466 of human vimentin using the numbering given in entry NP_003371.2 (GeneID 7431).

Biochem/physiol Actions

Vimentin helps to preserve the structure of cell and stability of tissue in mesenchymal cells. It is involved in tumorigenesis, EMT (epithelial mesenchymal transition) and the metastatic spread of cancer. It also controls infectious internalization of human papillomavirus 16 pseudovirions.

Physical form

Tris-buffered Saline containing 0.1% BSA containing 0.09% Sodium Azide

Other Notes

Vimentin is an intermediate filament expressed mainly in mesenchymal tissue and in primitive (undifferentiated) non-mesenchymal cells. It is a Class III cytoplasmic filament that forms a dense cytoskeletal network that extends from the nucleus to the plasma membrane. Vimentin reorganization during mitosis is likely regulated via its multiple phosphorylation sites. Vimentin expression is also found in mesoderm derived epithelia. Immunohistochemistry (IHC) with anti-vimentin antibodies is often used to identify neoplasms of mesenchymal or mesodermal origin.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vimentin modulates infectious internalisation of HPV16 pseudovirions
Schafer G, et al.
Journal of Virology, JVI-00307 (2017)
Mohammad Mehedi Hasan et al.
International journal of molecular sciences, 21(15) (2020-08-01)
While follicular fluid (FF) is well known to provide an optimal environment for oogenesis, its functional roles following its release into the oviduct during ovulation are currently elusive. We hypothesized that FF and FF-derived extracellular vesicles (EVs) may be conveyors
Qurat Ul Ain Reshi et al.
Journal of cell communication and signaling, 14(4), 439-451 (2020-09-04)
The capability of spermatozoa to directly influence maternal gene expression is already established. Indeed, some of the changes induced by spermatozoa may have a direct functional importance in the pre-conceptional period. Although the mechanisms underlying these sperm-maternal interactions are not
Viscoelastic properties of vimentin compared with other filamentous biopolymer networks.
Janmey PA, et al.
The Journal of Cell Biology, 113(1), 155-160 (1991)
Arun Satelli et al.
Cellular and molecular life sciences : CMLS, 68(18), 3033-3046 (2011-06-04)
Vimentin, a major constituent of the intermediate filament family of proteins, is ubiquitously expressed in normal mesenchymal cells and is known to maintain cellular integrity and provide resistance against stress. Vimentin is overexpressed in various epithelial cancers, including prostate cancer

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service