Skip to Content
Merck
All Photos(1)

Key Documents

P8102

Sigma-Aldrich

PAPA NONOate

Synonym(s):

3-(2-Hydroxy-2-nitroso-1-propylhydrazino)-1-propanamine, NOC-15

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H16N4O2
CAS Number:
Molecular Weight:
176.22
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Pricing and availability is not currently available.

shipped in

dry ice

storage temp.

−70°C

SMILES string

CCCN(CCCCN)N(O)N=O

General description

NO donor

Biochem/physiol Actions

NO release: t1/2 = 77 min (phosphate buffer at 22 °C)

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C M Maragos et al.
Cancer research, 53(3), 564-568 (1993-02-01)
Cell-mediated antitumor effects have, in part, been attributed to the production of NO. Compounds which generate NO might, therefore, be useful in attenuating the growth of tumor cells. Six nitric oxide/nucleophile adducts that release NO spontaneously in solution were tested
Hrabie, J.A., et al.
The Journal of Organic Chemistry, 58, 1472-1472 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service