Skip to Content
Merck
All Photos(1)

Key Documents

P1679

Sigma-Aldrich

Potassium tetrachloroplatinate(II)

Synonym(s):

Potassium platinum(II) chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2PtCl4
CAS Number:
Molecular Weight:
415.09
EC Number:
MDL number:
UNSPSC Code:
12352200

density

3.38 g/mL at 25 °C (lit.)

SMILES string

[K+].[K+].Cl[Pt--](Cl)(Cl)Cl

InChI

1S/4ClH.2K.Pt/h4*1H;;;/q;;;;2*+1;+2/p-4

InChI key

RVRDLMCWUILSAH-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Reacts with bis(dithiolates) to form metal-bis(dithiolates) with applications in laser Q-switch materials, liquid crystals, optical CD recording media, bar code materials and superconductivity. Reacts with capping polymers to form shape-control colloidal platinum nanoparticles for catalysis and photocatalysts.

replaced by

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service