Skip to Content
Merck
All Photos(1)

Key Documents

P0679

Sigma-Aldrich

Phosphorus pentoxide

ACS reagent, ≥98%

Synonym(s):

Phosphoric anhydride, Phosphorus(V) oxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
P2O5
CAS Number:
Molecular Weight:
141.94
EC Number:
UNSPSC Code:
12352300

grade

ACS reagent

vapor density

4.9 (vs air)

vapor pressure

1 mmHg ( 384 °C)
10 mmHg ( 238 °C)

Assay

≥98%

impurities

Phosphorus trioxide (P2O3), passes test (limit 0.02%)
≤0.02% Insoluble matter

mp

340 °C (lit.)

density

2.3 g/mL at 25 °C (lit.)

cation traces

NH4+: ≤0.01%
heavy metals (as Pb): ≤0.01%

SMILES string

O=P12OP3(=O)OP(=O)(O1)OP(=O)(O2)O3

InChI

1S/O10P4/c1-11-5-12(2)8-13(3,6-11)10-14(4,7-11)9-12

InChI key

YWEUIGNSBFLMFL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1A

Supplementary Hazards

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service